Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACCAATGACCACGAGCAGGA[C/T]ACCGATGACCACCTGCAGGAGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612220 MIM: 607297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALNT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALNT3 - beta-1,4-N-acetyl-galactosaminyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NINJ2 - ninjurin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294345.1 | 625 | Missense Mutation | ATC,GTC | I,V 75 | NP_001281274.1 | |
NM_001294346.1 | 625 | Missense Mutation | ATC,GTC | I,V 46 | NP_001281275.1 | |
NM_016533.5 | 625 | Missense Mutation | ATC,GTC | I,V 128 | NP_057617.2 |