Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCGCCGCCCGGACTCCCGACTC[A/G]GAAGTCGCGCCCGCCGCTCCGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRASP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRASP - general receptor for phosphoinositides 1 associated scaffold protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271856.1 | 159 | Intron | NP_001258785.1 | |||
NM_181711.3 | 159 | Silent Mutation | TCA,TCG | S,S 28 | NP_859062.1 | |
XM_005268691.3 | 159 | Intron | XP_005268748.1 | |||
XM_011537996.1 | 159 | Intron | XP_011536298.1 |