Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAATGCAAACCGATGCAACGCAC[A/G]GGTGTCAGGCTGTAACCCGGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605613 MIM: 610037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIP1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIP1R - huntingtin interacting protein 1 related | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS37B - VPS37B, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024667.2 | 1176 | UTR 3 | NP_078943.1 | |||
XM_005253622.3 | 1176 | UTR 3 | XP_005253679.1 | |||
XM_006719602.3 | 1176 | UTR 3 | XP_006719665.1 | |||
XM_011538743.2 | 1176 | UTR 3 | XP_011537045.1 | |||
XM_011538745.2 | 1176 | UTR 3 | XP_011537047.1 | |||
XM_017019970.1 | 1176 | Intron | XP_016875459.1 | |||
XM_017019971.1 | 1176 | Intron | XP_016875460.1 | |||
XM_017019972.1 | 1176 | Intron | XP_016875461.1 |