Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATGCACTCAAATGCAAACATAT[C/T]TGAAGAGTTTGAAATGTAAACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185605 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT1 - synaptotagmin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135805.1 | Intron | NP_001129277.1 | ||||
NM_001135806.1 | Intron | NP_001129278.1 | ||||
NM_001291901.1 | Intron | NP_001278830.1 | ||||
NM_005639.2 | Intron | NP_005630.1 | ||||
XM_005269113.2 | Intron | XP_005269170.1 | ||||
XM_006719576.1 | Intron | XP_006719639.1 | ||||
XM_011538710.1 | Intron | XP_011537012.1 | ||||
XM_017019909.1 | Intron | XP_016875398.1 |