Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGGTTCTGCTCCTCCTCGAAAG[C/T]CTTGAACAGTGAGTTGAAGTTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609695 MIM: 611055 | ||||||||||||||||||||
Literature Links: |
HPD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPD - 4-hydroxyphenylpyruvate dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171993.1 | 1148 | Missense Mutation | ACT,GCT | T,A 331 | NP_001165464.1 | |
NM_002150.2 | 1148 | Missense Mutation | ACT,GCT | T,A 370 | NP_002141.1 |
SETD1B - SET domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |