Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAGGGCTCCCTGAATCACTCAC[A/G]TGCGCACGAGCTTTAACTTCTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608352 MIM: 608524 | ||||||||||||||||||||
Literature Links: |
ACRBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACRBP - acrosin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ING4 - inhibitor of growth family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127582.1 | 538 | Missense Mutation | ACA,ATA | T,I 167 | NP_001121054.1 | |
NM_001127583.1 | 538 | Missense Mutation | ACA,ATA | T,I 164 | NP_001121055.1 | |
NM_001127584.1 | 538 | Missense Mutation | ACA,ATA | T,I 163 | NP_001121056.1 | |
NM_001127585.1 | 538 | Missense Mutation | ACA,ATA | T,I 143 | NP_001121057.1 | |
NM_001127586.1 | 538 | Missense Mutation | ACT,ATT | T,I 167 | NP_001121058.1 | |
NM_016162.3 | 538 | Missense Mutation | ACA,ATA | T,I 166 | NP_057246.2 | |
XM_005253698.3 | 538 | Missense Mutation | ACA,ATA | T,I 118 | XP_005253755.1 | |
XM_011520964.2 | 538 | Missense Mutation | ACA,ATA | T,I 142 | XP_011519266.1 | |
XM_011520965.2 | 538 | Missense Mutation | ACA,ATA | T,I 140 | XP_011519267.1 | |
XM_017019392.1 | 538 | Missense Mutation | ACA,ATA | T,I 117 | XP_016874881.1 | |
XM_017019393.1 | 538 | Missense Mutation | ACA,ATA | T,I 114 | XP_016874882.1 |