Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAATTTTTACTTTAATAACCAT[A/G]AAACTGATTTTTCACCTTCATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf73 - chromosome 12 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135570.1 | 1183 | UTR 3 | NP_001129042.1 | |||
XM_011538718.2 | 1183 | UTR 3 | XP_011537020.1 | |||
XM_017019916.1 | 1183 | UTR 3 | XP_016875405.1 | |||
XM_017019917.1 | 1183 | UTR 3 | XP_016875406.1 |
HSP90B1 - heat shock protein 90 beta family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |