Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTACTGGGAGGAGGAGCTGGTGCC[C/T]GATGCAGCCACACTGTCCGGAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609429 | ||||||||||||||||||||
Literature Links: |
FOXN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXN4 - forkhead box N4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213596.2 | 1785 | Silent Mutation | TCA,TCG | S,S 497 | NP_998761.2 | |
XM_011537922.2 | 1785 | Silent Mutation | TCA,TCG | S,S 498 | XP_011536224.1 | |
XM_011537923.2 | 1785 | Intron | XP_011536225.1 | |||
XM_017018818.1 | 1785 | Silent Mutation | TCA,TCG | S,S 454 | XP_016874307.1 | |
XM_017018819.1 | 1785 | Intron | XP_016874308.1 |