Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGCTTGAAGGACCCCATTACC[G/T]CCTCCTTGGAAAACCTCTCCGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160781 | ||||||||||||||||||||
Literature Links: |
CCDC63 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC63 - coiled-coil domain containing 63 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL2 - myosin light chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000432.3 | 472 | Missense Mutation | GAG,GCG | E,A 134 | NP_000423.2 |