Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTGTCCGTGCGGAAGGGGAG[G/T]CTTGAGGGGGACCAGGAAGGGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CSRNP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSRNP2 - cysteine and serine rich nuclear protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030809.2 | 1748 | Missense Mutation | AGA,AGC | R,S 507 | NP_110436.1 | |
XM_006719621.2 | 1748 | Missense Mutation | AGA,AGC | R,S 507 | XP_006719684.1 | |
XM_017019991.1 | 1748 | Missense Mutation | AGA,AGC | R,S 268 | XP_016875480.1 |