Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCAGGTGATGAAAAGCAAAAAG[C/G]AACCAATCTCTCTGTGGTGTTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTG1 - BTG anti-proliferation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001731.2 | 2287 | UTR 3 | NP_001722.1 |
LINC01619 - long intergenic non-protein coding RNA 1619 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928617 - uncharacterized LOC101928617 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |