Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCCCTGGAGCTTGAGGATAAG[A/G]TCAAGGTTGGTATGTGTTGCTCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC92 - coiled-coil domain containing 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304957.1 | 1792 | UTR 3 | NP_001291886.1 | |||
NM_001304958.1 | 1792 | UTR 3 | NP_001291887.1 | |||
NM_001304959.1 | 1792 | UTR 3 | NP_001291888.1 | |||
NM_001304960.1 | 1792 | UTR 3 | NP_001291889.1 | |||
NM_001304961.1 | 1792 | UTR 3 | NP_001291890.1 | |||
NM_025140.2 | 1792 | UTR 3 | NP_079416.1 | |||
XM_005253624.2 | 1792 | UTR 3 | XP_005253681.1 | |||
XM_017019984.1 | 1792 | UTR 3 | XP_016875473.1 |
DNAH10 - dynein axonemal heavy chain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207437.3 | 1792 | Intron | NP_997320.2 | |||
XM_005253555.3 | 1792 | Intron | XP_005253612.1 | |||
XM_011538014.2 | 1792 | Intron | XP_011536316.1 | |||
XM_011538015.2 | 1792 | Intron | XP_011536317.1 | |||
XM_011538016.2 | 1792 | Intron | XP_011536318.1 | |||
XM_011538017.2 | 1792 | Intron | XP_011536319.1 | |||
XM_011538019.2 | 1792 | Intron | XP_011536321.1 | |||
XM_011538020.2 | 1792 | Intron | XP_011536322.1 | |||
XM_017018960.1 | 1792 | Intron | XP_016874449.1 | |||
XM_017018961.1 | 1792 | Intron | XP_016874450.1 | |||
XM_017018962.1 | 1792 | Intron | XP_016874451.1 |