Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGGTAACGTTTTTTATTTCAC[A/G]TTCATTTTATTATTTCTGATGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC17 - zinc finger DHHC-type containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015336.2 | Intron | NP_056151.2 | ||||
XM_005268750.4 | Intron | XP_005268807.1 | ||||
XM_011538083.1 | Intron | XP_011536385.1 | ||||
XM_017019091.1 | Intron | XP_016874580.1 | ||||
XM_017019092.1 | Intron | XP_016874581.1 | ||||
XM_017019093.1 | Intron | XP_016874582.1 | ||||
XM_017019094.1 | Intron | XP_016874583.1 | ||||
XM_017019095.1 | Intron | XP_016874584.1 |