Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGGATTTTTAAGATGACCAAG[C/G]TTCCAGCCACCAAGAAGCTTCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC60 - coiled-coil domain containing 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178499.4 | 258 | Missense Mutation | CTT,GTT | L,V 4 | NP_848594.2 | |
XM_017018911.1 | 258 | Missense Mutation | CTT,GTT | L,V 4 | XP_016874400.1 | |
XM_017018912.1 | 258 | Missense Mutation | CTT,GTT | L,V 4 | XP_016874401.1 | |
XM_017018913.1 | 258 | Missense Mutation | CTT,GTT | L,V 4 | XP_016874402.1 | |
XM_017018914.1 | 258 | Intron | XP_016874403.1 | |||
XM_017018915.1 | 258 | Intron | XP_016874404.1 | |||
XM_017018916.1 | 258 | Intron | XP_016874405.1 |