Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGGGAGTCGCTGCCTGTGCCG[C/G]TGCACCCGCAGGTGCTGGGCGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DDX55 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX55 - DEAD-box helicase 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020936.2 | 59 | Missense Mutation | CTG,GTG | L,V 16 | NP_065987.1 | |
XM_011538608.2 | 59 | Missense Mutation | CTG,GTG | L,V 16 | XP_011536910.1 | |
XM_017019710.1 | 59 | Missense Mutation | CTG,GTG | L,V 16 | XP_016875199.1 | |
XM_017019711.1 | 59 | UTR 5 | XP_016875200.1 | |||
XM_017019712.1 | 59 | Intron | XP_016875201.1 | |||
XM_017019713.1 | 59 | UTR 5 | XP_016875202.1 | |||
XM_017019714.1 | 59 | UTR 5 | XP_016875203.1 | |||
XM_017019715.1 | 59 | Missense Mutation | CTG,GTG | L,V 16 | XP_016875204.1 | |
XM_017019716.1 | 59 | Missense Mutation | CTG,GTG | L,V 16 | XP_016875205.1 | |
XM_017019717.1 | 59 | UTR 5 | XP_016875206.1 | |||
XM_017019718.1 | 59 | Intron | XP_016875207.1 | |||
XM_017019719.1 | 59 | UTR 5 | XP_016875208.1 | |||
XM_017019720.1 | 59 | UTR 5 | XP_016875209.1 | |||
XM_017019721.1 | 59 | Missense Mutation | CTG,GTG | L,V 16 | XP_016875210.1 |
TMED2 - transmembrane p24 trafficking protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |