Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCAAGTTTCCAGCCGGGAAGAA[G/T]CCTCTCAGCCGTAGGCGTCTTTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM101A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM101A - family with sequence similarity 101 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181709.4 | 170 | UTR 5 | NP_859060.3 |
LOC105370045 - filamin-interacting protein FAM101A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF664-FAM101A - filamin-interacting protein FAM101A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204299.1 | 170 | Intron | NP_001191228.1 |