Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAATCCTTGCTGAGTGGGGAGG[C/T]CCACGCGGGGGGGCAGGGGCGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605613 MIM: 610037 | ||||||||||||||||||||
Literature Links: |
HIP1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIP1R - huntingtin interacting protein 1 related | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS37B - VPS37B, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024667.2 | 846 | Missense Mutation | GAC,GGC | D,G 248 | NP_078943.1 | |
XM_005253622.3 | 846 | Missense Mutation | GAC,GGC | D,G 220 | XP_005253679.1 | |
XM_006719602.3 | 846 | Missense Mutation | GAC,GGC | D,G 220 | XP_006719665.1 | |
XM_011538743.2 | 846 | Missense Mutation | GAC,GGC | D,G 307 | XP_011537045.1 | |
XM_011538745.2 | 846 | Missense Mutation | GAC,GGC | D,G 230 | XP_011537047.1 | |
XM_017019970.1 | 846 | Intron | XP_016875459.1 | |||
XM_017019971.1 | 846 | Intron | XP_016875460.1 | |||
XM_017019972.1 | 846 | Intron | XP_016875461.1 |