Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGAATTCTGCCTAGCCAGAGAC[C/T]CACACCTCCTTGTGATAACGGACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604874 | ||||||||||||||||||||
Literature Links: |
KLRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLRG1 - killer cell lectin like receptor G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005810.3 | 878 | Missense Mutation | CCA,TCA | P,S 108 | NP_005801.3 | |
XM_017018682.1 | 878 | Missense Mutation | CCA,TCA | P,S 108 | XP_016874171.1 | |
XM_017018683.1 | 878 | Missense Mutation | CCA,TCA | P,S 108 | XP_016874172.1 | |
XM_017018684.1 | 878 | Missense Mutation | CCA,TCA | P,S 108 | XP_016874173.1 | |
XM_017018685.1 | 878 | Missense Mutation | CCA,TCA | P,S 108 | XP_016874174.1 |