Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGGAGCTGTTTCCAGTCCCCCC[A/G]TCTCCAGTGGGTGTGGAGAACAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611181 MIM: 604986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACAD10 - acyl-CoA dehydrogenase family member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136538.1 | 232 | Missense Mutation | CAT,CGT | H,R 11 | NP_001130010.1 | |
NM_025247.5 | 232 | Missense Mutation | CAT,CGT | H,R 11 | NP_079523.3 |
BRAP - BRCA1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |