Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCAGGGCTCGCAGAAGGCTCTCC[A/G]TACGGGGCTGCAGCCTGTGGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DHX37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX37 - DEAH-box helicase 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032656.3 | 3408 | Missense Mutation | ACG,ATG | T,M 1102 | NP_116045.2 | |
XM_005253590.3 | 3408 | Missense Mutation | ACG,ATG | T,M 1102 | XP_005253647.1 | |
XM_011538598.2 | 3408 | Intron | XP_011536900.1 | |||
XM_011538600.2 | 3408 | Intron | XP_011536902.1 |