Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAAACAAACCAGGCCCAACACT[C/G]TGTGTTTAGAAAGAGAATTTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613397 MIM: 608711 MIM: 613057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVIL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AVIL - advillin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CTDSP2 - CTD small phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005730.3 | 2148 | UTR 3 | NP_005721.3 | |||
XM_005268556.1 | 2148 | UTR 3 | XP_005268613.1 |
MIR26A2 - microRNA 26a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |