Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATCTTACCTGGTTTTTATTTGG[A/G]TTCTCCATGAAGACACACTGCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD5 - C2 calcium dependent domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286173.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1005 | NP_001273102.1 | |
NM_001286174.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1003 | NP_001273103.1 | |
NM_001286175.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1006 | NP_001273104.1 | |
NM_001286176.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1003 | NP_001273105.1 | |
NM_001286177.1 | 2280 | Silent Mutation | AAC,AAT | N,N 994 | NP_001273106.1 | |
NM_014802.2 | 2280 | Silent Mutation | AAC,AAT | N,N 952 | NP_055617.1 | |
XM_005253538.2 | 2280 | Silent Mutation | AAC,AAT | N,N 943 | XP_005253595.1 | |
XM_005253539.3 | 2280 | Silent Mutation | AAC,AAT | N,N 625 | XP_005253596.1 | |
XM_006719170.2 | 2280 | Silent Mutation | AAC,AAT | N,N 969 | XP_006719233.1 | |
XM_017020264.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1113 | XP_016875753.1 | |
XM_017020265.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1103 | XP_016875754.1 | |
XM_017020266.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1096 | XP_016875755.1 | |
XM_017020267.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1089 | XP_016875756.1 | |
XM_017020268.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1067 | XP_016875757.1 | |
XM_017020269.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1062 | XP_016875758.1 | |
XM_017020270.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1049 | XP_016875759.1 | |
XM_017020271.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1040 | XP_016875760.1 | |
XM_017020272.1 | 2280 | Silent Mutation | AAC,AAT | N,N 1016 | XP_016875761.1 | |
XM_017020273.1 | 2280 | Silent Mutation | AAC,AAT | N,N 999 | XP_016875762.1 | |
XM_017020274.1 | 2280 | Silent Mutation | AAC,AAT | N,N 993 | XP_016875763.1 | |
XM_017020275.1 | 2280 | Silent Mutation | AAC,AAT | N,N 989 | XP_016875764.1 | |
XM_017020276.1 | 2280 | Silent Mutation | AAC,AAT | N,N 984 | XP_016875765.1 | |
XM_017020277.1 | 2280 | Silent Mutation | AAC,AAT | N,N 977 | XP_016875766.1 | |
XM_017020278.1 | 2280 | Silent Mutation | AAC,AAT | N,N 965 | XP_016875767.1 | |
XM_017020279.1 | 2280 | Silent Mutation | AAC,AAT | N,N 731 | XP_016875768.1 |