Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACAAGGGGGAGTCCTGCGGCGG[C/T]GGTCGAGGGGAGCTGGCTGAGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000545.6 | 357 | Silent Mutation | GGC,GGT | G,G 52 | NP_000536.5 | |
NM_001306179.1 | 357 | Silent Mutation | GGC,GGT | G,G 52 | NP_001293108.1 | |
XM_005253931.3 | 357 | Silent Mutation | GGC,GGT | G,G 52 | XP_005253988.1 |
HNF1A-AS1 - HNF1A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |