Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGAAGAAGGAGACCGGAGCCGCC[G/T]CCTTCCTCTGCCGCTACCCGGAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190470 | ||||||||||||||||||||
Literature Links: |
TPP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPP2 - tripeptidyl peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005254070.1 | 175 | Missense Mutation | GCC,TCC | A,S 25 | XP_005254127.1 | |
XM_005254071.1 | 175 | Missense Mutation | GCC,TCC | A,S 25 | XP_005254128.1 | |
XM_005254072.1 | 175 | Missense Mutation | GCC,TCC | A,S 25 | XP_005254129.1 |