Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGAGTGTTGAATTTCATGAAA[A/G]AGGAAAGAATCATAAGGAAAATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189973 MIM: 604981 | ||||||||||||||||||||
Literature Links: |
ELF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELF1 - E74 like ETS transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP4 - WW domain binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007187.3 | 185 | Missense Mutation | AAA,AGA | K,R 32 | NP_009118.1 | |
XM_005266245.2 | 185 | Missense Mutation | AAA,AGA | K,R 63 | XP_005266302.1 | |
XM_017020380.1 | 185 | UTR 5 | XP_016875869.1 |