Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATAAAGCTAATTACTAACATG[A/G]CCTAAGACAGAACCTAAGCTGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610793 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A30 - solute carrier family 25 member 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010875.3 | 1974 | UTR 3 | NP_001010875.1 | |||
NM_001286806.1 | 1974 | UTR 3 | NP_001273735.1 | |||
NM_001286807.1 | 1974 | UTR 3 | NP_001273736.1 | |||
XM_005266321.2 | 1974 | UTR 3 | XP_005266378.1 | |||
XM_005266322.1 | 1974 | Intron | XP_005266379.1 | |||
XM_006719793.3 | 1974 | Intron | XP_006719856.1 | |||
XM_006719794.1 | 1974 | Intron | XP_006719857.1 | |||
XM_011535026.1 | 1974 | Intron | XP_011533328.1 | |||
XM_011535027.2 | 1974 | UTR 3 | XP_011533329.1 | |||
XM_017020523.1 | 1974 | Intron | XP_016876012.1 | |||
XM_017020524.1 | 1974 | UTR 3 | XP_016876013.1 |
TPT1-AS1 - TPT1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |