Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAATTTCACGAGAAATGCAACAC[A/G]TTACAGCGTCCAGCTCATGAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAMP1 - lysosomal associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005561.3 | 610 | Missense Mutation | CAT,CGT | H,R 110 | NP_005552.3 | |
XM_011537494.1 | 610 | Missense Mutation | CAT,CGT | H,R 91 | XP_011535796.1 |