Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTTTGCTTGCTCAAGGTTCCC[A/G]CTTACTTTGCATCTCACTGTTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606922 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00359 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00359 - long intergenic non-protein coding RNA 359 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OXGR1 - oxoglutarate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080818.3 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | NP_543008.3 | |
XM_005254043.3 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | XP_005254100.1 | |
XM_005254044.3 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | XP_005254101.1 | |
XM_005254045.2 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | XP_005254102.1 | |
XM_005254046.2 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | XP_005254103.1 | |
XM_011521077.2 | 1347 | Silent Mutation | AGC,AGT | S,S 322 | XP_011519379.1 |