Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAAAACTCTTCTGCATATTCAC[C/T]ATAACGATTATCTGCCGGGCTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603561 | ||||||||||||||||||||
Literature Links: |
MTMR6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTMR6 - myotubularin related protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004685.3 | 1370 | Missense Mutation | AGT,GGT | S,G 596 | NP_004676.3 | |
XM_011535306.1 | 1370 | Missense Mutation | AGT,GGT | S,G 634 | XP_011533608.1 | |
XM_011535307.1 | 1370 | UTR 3 | XP_011533609.1 | |||
XM_017020845.1 | 1370 | UTR 3 | XP_016876334.1 | |||
XM_017020846.1 | 1370 | Missense Mutation | AGT,GGT | S,G 401 | XP_016876335.1 |