Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCACCTTACAGGTCTGGGATGTA[C/T]CTTTCCATCTGTTGCTGCTTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611760 | ||||||||||||||||||||
Literature Links: |
PCDH17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDH17 - protocadherin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040429.2 | 2449 | Silent Mutation | TAC,TAT | Y,Y 2 | NP_001035519.1 | |
XM_005266357.2 | 2449 | Silent Mutation | TAC,TAT | Y,Y 2 | XP_005266414.1 | |
XM_005266358.2 | 2449 | Silent Mutation | TAC,TAT | Y,Y 2 | XP_005266415.1 | |
XM_017020547.1 | 2449 | Silent Mutation | TAC,TAT | Y,Y 2 | XP_016876036.1 |