Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGCGCCGGGCCTGGTGCTCTG[A/C]GATCAAGGGCTGCAGAACCTCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609911 MIM: 191050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A47 - solute carrier family 25 member 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004184.3 | 1682 | Missense Mutation | NP_004175.2 | |||
NM_173701.1 | 1682 | Missense Mutation | NP_776049.1 | |||
NM_213645.1 | 1682 | Missense Mutation | NP_998810.1 | |||
NM_213646.1 | 1682 | Missense Mutation | NP_998811.1 | |||
XM_005268044.3 | 1682 | Missense Mutation | XP_005268101.1 | |||
XM_006720249.3 | 1682 | Missense Mutation | XP_006720312.1 | |||
XM_011537133.2 | 1682 | Missense Mutation | XP_011535435.1 | |||
XM_011537135.2 | 1682 | Missense Mutation | XP_011535437.1 | |||
XM_011537136.2 | 1682 | Missense Mutation | XP_011535438.1 | |||
XM_017021627.1 | 1682 | Missense Mutation | XP_016877116.1 | |||
XM_017021628.1 | 1682 | Missense Mutation | XP_016877117.1 | |||
XM_017021629.1 | 1682 | Missense Mutation | XP_016877118.1 |