Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGAAGAAAGCATGGCGTGAGT[A/G]CTCTAAGGATAGACCTACGGTATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP36L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP36L1 - ZFP36 ring finger protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244698.1 | 2334 | UTR 3 | NP_001231627.1 | |||
NM_001244701.1 | 2334 | UTR 3 | NP_001231630.1 | |||
NM_004926.3 | 2334 | UTR 3 | NP_004917.2 |