Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTGTCTGTTCCTCTTCGTTTAA[C/T]CCCTCATCTCCTTCCAGAGGGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004184.3 | Intron | NP_004175.2 | ||||
NM_173701.1 | Intron | NP_776049.1 | ||||
NM_213645.1 | Intron | NP_998810.1 | ||||
NM_213646.1 | Intron | NP_998811.1 | ||||
XM_005268044.3 | Intron | XP_005268101.1 | ||||
XM_006720249.3 | Intron | XP_006720312.1 | ||||
XM_011537133.2 | Intron | XP_011535435.1 | ||||
XM_011537135.2 | Intron | XP_011535437.1 | ||||
XM_011537136.2 | Intron | XP_011535438.1 | ||||
XM_017021627.1 | Intron | XP_016877116.1 | ||||
XM_017021628.1 | Intron | XP_016877117.1 | ||||
XM_017021629.1 | Intron | XP_016877118.1 |
WDR25 - WD repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161476.1 | Intron | NP_001154948.1 | ||||
NM_024515.4 | Intron | NP_078791.3 | ||||
XM_005268056.4 | Intron | XP_005268113.1 | ||||
XM_005268057.3 | Intron | XP_005268114.1 | ||||
XM_005268058.4 | Intron | XP_005268115.1 | ||||
XM_011537150.2 | Intron | XP_011535452.1 | ||||
XM_011537151.2 | Intron | XP_011535453.2 | ||||
XM_017021639.1 | Intron | XP_016877128.1 |