Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGCCTTGCCCATCCTCGGTGAT[A/G]TCTAGGGTTGTCACTCTGGTGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC85C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC85C - coiled-coil domain containing 85C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HHIPL1 - HHIP like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006720277.3 | Intron | XP_006720340.1 | ||||
XM_011537236.2 | Intron | XP_011535538.1 | ||||
XM_011537237.2 | Intron | XP_011535539.1 | ||||
XM_017021707.1 | Intron | XP_016877196.1 | ||||
XM_017021708.1 | Intron | XP_016877197.1 |