Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAATTTACTTTGTTAAAGGTCTC[A/C]AGATTCTGCCCTCCAAGAAAGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616003 MIM: 603885 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOPT1 - apoptogenic 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302652.1 | 174 | Silent Mutation | TCA,TCC | S,S 56 | NP_001289581.1 | |
NM_001302653.1 | 174 | Silent Mutation | TCA,TCC | S,S 56 | NP_001289582.1 | |
NM_001302654.1 | 174 | Silent Mutation | TCA,TCC | S,S 56 | NP_001289583.1 | |
NM_032374.4 | 174 | Silent Mutation | TCA,TCC | S,S 56 | NP_115750.2 |
BAG5 - BCL2 associated athanogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |