Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAAGAAAAGCTGAAAGGGAGAC[A/G]TTTGCAGTGAGTGGTTAGAGCAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600754 MIM: 611856 | ||||||||||||||||||||
Literature Links: |
MMP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP14 - matrix metallopeptidase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL52 - mitochondrial ribosomal protein L52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318870.1 | 329 | Silent Mutation | ACA,ACG | T,T 71 | NP_001305799.1 | |
NM_178336.2 | 329 | Silent Mutation | ACA,ACG | T,T 72 | NP_848026.1 | |
NM_180982.2 | 329 | Silent Mutation | ACA,ACG | T,T 71 | NP_851313.1 | |
NM_181304.2 | 329 | Silent Mutation | ACA,ACG | T,T 13 | NP_851821.1 | |
NM_181305.2 | 329 | Silent Mutation | ACA,ACG | T,T 13 | NP_851822.1 | |
NM_181306.2 | 329 | Silent Mutation | ACA,ACG | T,T 13 | NP_851823.1 | |
NM_181307.2 | 329 | Intron | NP_851824.1 | |||
XM_005267325.4 | 329 | Silent Mutation | ACA,ACG | T,T 72 | XP_005267382.1 | |
XM_005267328.3 | 329 | Silent Mutation | ACA,ACG | T,T 13 | XP_005267385.1 |