Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACAGTCCATACGTACACTGTCC[C/T]ATCTCAATTCTACATGTAGTCCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDHD1 - DDHD domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160147.1 | 9754 | UTR 3 | NP_001153619.1 | |||
NM_001160148.1 | 9754 | UTR 3 | NP_001153620.1 | |||
NM_030637.2 | 9754 | UTR 3 | NP_085140.2 | |||
XM_005268102.2 | 9754 | Intron | XP_005268159.1 | |||
XM_005268103.2 | 9754 | Intron | XP_005268160.1 | |||
XM_005268105.2 | 9754 | Intron | XP_005268162.1 | |||
XM_011537188.2 | 9754 | UTR 3 | XP_011535490.1 | |||
XM_011537189.2 | 9754 | Intron | XP_011535491.1 | |||
XM_017021668.1 | 9754 | Intron | XP_016877157.1 | |||
XM_017021669.1 | 9754 | Intron | XP_016877158.1 |