Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGTTTCTGGTAACGCTTTATGA[A/G]TACTCGCCGCTTTTCTACATCGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606138 | ||||||||||||||||||||
Literature Links: |
CGRRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CGRRF1 - cell growth regulator with ring finger domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006568.2 | 97 | Silent Mutation | GAA,GAG | E,E 11 | NP_006559.1 | |
XM_011536362.2 | 97 | Intron | XP_011534664.1 | |||
XM_017020931.1 | 97 | UTR 5 | XP_016876420.1 | |||
XM_017020932.1 | 97 | Intron | XP_016876421.1 |