Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTCACCTCAGCTTAAGGATCA[C/T]AGCACCGCACTTGACTTCCAGGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613431 | ||||||||||||||||||||
Literature Links: |
HAUS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAUS4 - HAUS augmin like complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166269.1 | Intron | NP_001159741.1 | ||||
NM_001166270.1 | Intron | NP_001159742.1 | ||||
NM_017815.2 | Intron | NP_060285.2 |
LOC101926933 - uncharacterized LOC101926933 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4707 - microRNA 4707 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |