Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGTATGATCTTAAGACTATATG[C/T]ACTGAGTCCTATTAGTCAGTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179617 MIM: 611873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD51 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD51 - RAD51 recombinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RMDN3 - regulator of microtubule dynamics 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304802.1 | 2172 | UTR 3 | NP_001291731.1 | |||
NM_001323894.1 | 2172 | UTR 3 | NP_001310823.1 | |||
NM_001323895.1 | 2172 | UTR 3 | NP_001310824.1 | |||
NM_001323896.1 | 2172 | UTR 3 | NP_001310825.1 | |||
NM_001323897.1 | 2172 | UTR 3 | NP_001310826.1 | |||
NM_018145.2 | 2172 | UTR 3 | NP_060615.1 | |||
XM_011521755.2 | 2172 | UTR 3 | XP_011520057.1 |