Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGTTGTGTTAGATGCACCACC[C/T]CGATGATCAGAGAGGGATTCAGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAGAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAGAB - alpha- and gamma-adaptin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271885.1 | 1024 | Silent Mutation | CGA,CGG | R,R 114 | NP_001258814.1 | |
NM_001271886.1 | 1024 | Silent Mutation | CGA,CGG | R,R 114 | NP_001258815.1 | |
NM_024666.4 | 1024 | Silent Mutation | CGA,CGG | R,R 223 | NP_078942.3 | |
XM_011522020.1 | 1024 | Silent Mutation | CGA,CGG | R,R 114 | XP_011520322.1 |