Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCAGCACCCAGGAGGCATGGCC[A/G]CTCAGCGTGCCAGCCAAATTGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR61 - WD repeat domain 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303247.1 | 891 | Silent Mutation | AGC,AGT | S,S 229 | NP_001290176.1 | |
NM_001303248.1 | 891 | Silent Mutation | AGC,AGT | S,S 136 | NP_001290177.1 | |
NM_025234.2 | 891 | Silent Mutation | AGC,AGT | S,S 229 | NP_079510.1 | |
XM_011522094.1 | 891 | Silent Mutation | AGC,AGT | S,S 229 | XP_011520396.1 | |
XM_017022637.1 | 891 | Silent Mutation | AGC,AGT | S,S 229 | XP_016878126.1 |