Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCTACATTGTCTTCCTCAGAA[A/C]CATTAAGGGTTTTTCCAAGCTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615756 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SECISBP2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SECISBP2L - SECIS binding protein 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193489.1 | 3380 | Missense Mutation | GGT,GTT | G,V 1039 | NP_001180418.1 | |
NM_014701.3 | 3380 | Missense Mutation | GGT,GTT | G,V 994 | NP_055516.2 |