Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCTTGGGAGGGGCTGCCTCAC[C/T]GGGGGCTGAGTGGTCCTTGAAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612864 | ||||||||||||||||||||
Literature Links: |
PLA2G4D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLA2G4D - phospholipase A2 group IVD | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178034.3 | 2325 | Missense Mutation | AGT,GGT | S,G 744 | NP_828848.3 | |
XM_011521467.2 | 2325 | Missense Mutation | AGT,GGT | S,G 688 | XP_011519769.1 |