Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGAGGAGATGATTGGCAAACTG[G/T]TATCCATATGGTATCTGATAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SHC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHC4 - SHC adaptor protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203349.3 | 2228 | Missense Mutation | AAC,ACC | N,T 600 | NP_976224.3 | |
XM_005254375.3 | 2228 | Missense Mutation | AAC,ACC | N,T 417 | XP_005254432.1 |