Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGCCAGGATCTGGAGTTTCAGGG[C/G]AGGCCTGGCAGTGCTGAGGCTGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616844 MIM: 613256 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC17 - DnaJ heat shock protein family (Hsp40) member C17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R14D - protein phosphatase 1 regulatory inhibitor subunit 14D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130143.1 | 549 | Silent Mutation | CTC,CTG | L,L 199 | NP_001123615.1 | |
NM_017726.7 | 549 | UTR 3 | NP_060196.1 | |||
XM_017022372.1 | 549 | Intron | XP_016877861.1 |
ZFYVE19 - zinc finger FYVE-type containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |