Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTGAAGAGGCCATGGGCTTAG[C/G]GGGGATGAGGTCCAAGTCCTCGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM219B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM219B - family with sequence similarity 219 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321920.1 | 611 | Missense Mutation | CCT,GCT | P,A 174 | NP_001308849.1 | |
NM_001321921.1 | 611 | Missense Mutation | CCT,GCT | P,A 174 | NP_001308850.1 | |
NM_001321922.1 | 611 | Missense Mutation | CCT,GCT | P,A 173 | NP_001308851.1 | |
NM_001321923.1 | 611 | Missense Mutation | CCT,GCT | P,A 173 | NP_001308852.1 | |
NM_020447.4 | 611 | Missense Mutation | CCT,GCT | P,A 174 | NP_065180.1 | |
XM_017022433.1 | 611 | Missense Mutation | CCT,GCT | P,A 173 | XP_016877922.1 |
MPI - mannose phosphate isomerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |