Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCCCTCTCATTTGTCCACAGTC[A/G]GACTATGGGGAAGAGGAGCTGTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS3 - ankyrin repeat and sterile alpha motif domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf71 - chromosome 16 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139170.2 | 323 | Silent Mutation | TCA,TCG | S,S 44 | NP_631909.2 | |
XM_005255144.3 | 323 | UTR 5 | XP_005255201.1 | |||
XM_017022975.1 | 323 | Silent Mutation | TCA,TCG | S,S 67 | XP_016878464.1 | |
XM_017022976.1 | 323 | Silent Mutation | TCA,TCG | S,S 44 | XP_016878465.1 |
ZNF500 - zinc finger protein 500 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |