Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACTGCGGTGTAGACAGCAGGGC[C/T]CCTGTGGGCATCTCAAAAGGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGAL - integrin subunit alpha L | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF747 - zinc finger protein 747 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF768 - zinc finger protein 768 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024671.3 | 664 | Silent Mutation | GGA,GGG | G,G 221 | NP_078947.3 | |
XM_017023665.1 | 664 | Silent Mutation | GGA,GGG | G,G 245 | XP_016879154.1 | |
XM_017023666.1 | 664 | Silent Mutation | GGA,GGG | G,G 190 | XP_016879155.1 |