Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCCATCACTGCTGATGATGCCAC[A/G]CATATGTGACCAGTCTGGGGGTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603365 MIM: 613199 | ||||||||||||||||||||
Literature Links: |
HIRIP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIRIP3 - HIRA interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197323.1 | 2127 | UTR 3 | NP_001184252.1 | |||
NM_003609.4 | 2127 | Missense Mutation | CGT,TGT | R,C 546 | NP_003600.2 |
INO80E - INO80 complex subunit E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAOK2 - TAO kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252043.1 | 2127 | Intron | NP_001238972.1 | |||
NM_004783.3 | 2127 | Intron | NP_004774.1 | |||
NM_016151.3 | 2127 | Intron | NP_057235.2 | |||
XM_011545982.1 | 2127 | Intron | XP_011544284.1 | |||
XM_011545983.2 | 2127 | Intron | XP_011544285.1 | |||
XM_011545984.1 | 2127 | Intron | XP_011544286.1 | |||
XM_011545985.1 | 2127 | Intron | XP_011544287.1 | |||
XM_011545986.2 | 2127 | Intron | XP_011544288.1 |